Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0008431M1

Sigma-Aldrich

Monoclonal Anti-NR0B2 antibody produced in mouse

clone 1A11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-SHP, Anti-SHP1, Anti-nuclear receptor subfamily 0, group B, member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR0B2(8431)

Related Categories

General description

The protein encoded by this gene is an unusual orphan receptor that contains a putative ligand-binding domain but lacks a conventional DNA-binding domain. The gene product is a member of the nuclear hormone receptor family, a group of transcription factors regulated by small hydrophobic hormones, a subset of which do not have known ligands and are referred to as orphan nuclear receptors. The protein has been shown to interact with retinoid and thyroid hormone receptors, inhibiting their ligand-dependent transcriptional activation. In addition, interaction with estrogen receptors has been demonstrated, leading to inhibition of function. Studies suggest that the protein represses nuclear hormone receptor-mediated transactivation via two separate steps: competition with coactivators and the direct effects of its transcriptional repressor function. (provided by RefSeq)

Immunogen

NR0B2 (AAH30207, 1 a.a. ~ 257 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSTSQPGACPCQGAASRPAILYALLSSSLKAVPRPRSRCLCRQHRPVQLCAPHRTCREALDVLAKTVAFLRNLPSFWQLPPQDQRRLLQGCWGPLFLLGLAQDAVTFEVAEAPVPSILKKILLEEPSSSGGSGQLPDRPQPSLAAVQWLQCCLESFWSLELSPKEYACLKGTILFNPDVPGLQAASHIGHLQQEAHWVLCEVLEPWCPAAQGRLTRVLLTASTLKSIPTSLLGDLFFRPIIGDVDIAGLLGDMLLLR

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service