Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0007267M9

Sigma-Aldrich

Monoclonal Anti-TTC3 antibody produced in mouse

clone 2D10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DCRR1, Anti-DKFZp686M0150, Anti-RNF105, Anti-TPRDIII, Anti-tetratricopeptide repeat domain 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$539.00

$539.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$539.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TTC3(7267)

General description

TTCR (tetratricopeptide repeat protein 3) is one of the main genes present within the Down syndrome critical region (DSCR) on human chromosome 21q22.2. The encoded protein is an E3 ubiquitin liase, composed of 2025 amino acids, and its N-terminal contains three tetratricopeptide repeat (TPR) motifs. It also contains a RING (really interesting new gene) finger motif, a putative Akt phosphorylation site, and nuclear localization signals.

Immunogen

TTC3 (NP_003307, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDNFAEGDFTVADYALLEDCPHVDDCVFAAEFMSNDYVRVTQLYCDGVGVQYKDYIQSERNLEFDICSIWCSKPISVLQDYCDAIKINIFWPLLFQHQNSSVISRLHPCV

Biochem/physiol Actions

TTCR (tetratricopeptide repeat protein 3) interacts with phosphorylated Akt protein, and promotes its ubiquitination and degradation in the nucleus. The expression of TTCR is increased in Down syndrome (DS) cells, and its interaction with Akt protein is thought to be responsible for the clinical symptoms of DS. TTC3-RhoA-CIT-K (citron kinase) pathway is thought to play a key role in neuronal development, and its hyperactivity might result in disruption of normal differentiation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaia Berto et al.
Journal of cell science, 120(Pt 11), 1859-1867 (2007-05-10)
The Down syndrome critical region (DSCR) on Chromosome 21 contains many genes whose duplication may lead to the major phenotypic features of Down syndrome and especially the associated mental retardation. However, the functions of DSCR genes are mostly unknown and
Futoshi Suizu et al.
Developmental cell, 17(6), 800-810 (2010-01-12)
The serine threonine kinase Akt is a core survival factor that underlies a variety of human diseases. Although regulatory phosphorylation and dephosphorylation have been well documented, the other posttranslational mechanisms that modulate Akt activity remain unclear. We show here that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service