Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0006928M1

Sigma-Aldrich

Monoclonal Anti-TCF2 antibody produced in mouse

clone 3H4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HNF1B, Anti-HNF1beta, Anti-HNF2, Anti-LFB3, Anti-MODY5, Anti-VHNF1, Anti-transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$539.00

$539.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1306


Select a Size

Change View
100 μG
$539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$539.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1306

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNF1B(6928)

General description

This gene encodes a member of the homeodomain-containing superfamily of transcription factors. The protein binds to DNA as either a homodimer, or a heterodimer with the related protein hepatocyte nuclear factor 1-alpha. The gene has been shown to function in nephron development, and regulates development of the embryonic pancreas. Mutations in this gene result in renal cysts and diabetes syndrome and noninsulin-dependent diabetes mellitus, and expression of this gene is altered in some types of cancer. Multiple transcript variants encoding different isoforms have been found for this gene

Immunogen

TCF2 (NP_000449, 29 a.a. ~ 118 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ALEELLPSPNFGVKLETLPLSPGSGAEPDTKPVFHTLTNGHAKGRLSGDEGSEDGDDYDTPPILKELQALNTEEAAEQRAEVDRMLSEDP

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service