Skip to Content
MilliporeSigma
All Photos(6)

Documents

WH0005873M2

Sigma-Aldrich

Monoclonal Anti-RAB27A antibody produced in mouse

clone 1G7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-GS2, Anti-HsT18676, Anti-MGC117246, Anti-RAB27, Anti-RAB27A, member RAS oncogene family, Anti-RAM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB27A(5873)

General description

The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. (provided by RefSeq)

Immunogen

RAB27A (NP_004571, 122 a.a. ~ 221 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YCENPDIVLCGNKSDLEDQRVVKEEEAIALAEKYGIPYFETSAANGTNISQAIEMLLDLIMKRMERCVDKSWIPEGVVRSNGHASTDQLSEEKEKGACGC

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAB27A/Melanophilin Blocker Inhibits Melanoma Cell Motility and Invasion.
Dajiang Guo et al.
The Journal of investigative dermatology, 140(7), 1470-1473 (2020-01-25)
Dajiang Guo et al.
Pigment cell & melanoma research, 33(6), 889-894 (2020-06-09)
The role of the small GTPase RAB27A as an essential melanosome trafficking regulator in melanocytes is well-accepted. A decade ago, RAB27A was identified as a tumor dependency gene that promotes melanoma cell proliferation. RAB27A has since been linked to another
Feixue Feng et al.
Oncology letters, 14(3), 2947-2953 (2017-09-21)
Cancer stem cells (CSCs) are associated with tumor initiation, therapeutic resistance, relapse and metastasis. However, the underlying mechanisms CSCs use to preserve stemness are not yet fully understood. The present study demonstrated that the expression of RAB27A, member RAS oncogene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service