Skip to Content
MilliporeSigma
All Photos(7)

Documents

WH0005238M1

Sigma-Aldrich

Monoclonal Anti-PGM3 antibody produced in mouse

clone 1E2-1B12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AGM1, Anti-DKFZp434B187, Anti-PAGM, Anti-phosphoglucomutase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E2-1B12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PGM3(5238)

Related Categories

Immunogen

PGM3 (AAH01258, 1 a.a. ~ 542 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDLGAITKYSALHAKPNGLILQYGTAGFRTKAEHLDHVMFRMGLLAVLRSKQTKSTIGVMVTASHNPEEDNGVKLVDPLGEMLAPSWEEHATCLANAEEQDMQRVLIDISEKEAVNLQQDAFVVIGRDTRPSSEKLSQSVIDGVTVLGGQFHDYGLLTTPQLHYMVYCRNTGGRYGKATIEGYYQKLSKAFVELTKQASCSGDEYRSLKVDCANGIGALKLREMEHYFSQGLSVQLFNDGSKGKLNHLCGADFVKSHQKPPQGMEIKSNERCCSFDGDADRIVYYYHDADGHFHLIDGDKIATLISSFLKELLVEIGESLNIGVVQTAYANGSSTRYLEEVMKVPVYCTKTGVKHLHHKAQEFDIGVYFEANGHGTALFSTAVEMKIKQSAEQLEDKKRKAAKMLENIIDLFNQAAGDAISDMLVIEAILALKGLTVQQWDALYTDLPNRQLKVQVADRRVISTTNAERQAVTPPGLQEAINDLVKKYKLSRAFVRPSGTEDVVRVYAEADSQESADHLAHEVSLAVFQLAGGIGERPQPGF

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Zhang et al.
The Journal of allergy and clinical immunology, 133(5), 1400-1409 (2014-03-05)
Identifying genetic syndromes that lead to significant atopic disease can open new pathways for investigation and intervention in allergy. We sought to define a genetic syndrome of severe atopy, increased serum IgE levels, immune deficiency, autoimmunity, and motor and neurocognitive
Diem Hong Tran et al.
Nature communications, 11(1), 1771-1771 (2020-04-15)
The hexosamine biosynthetic pathway (HBP) plays critical roles in nutrient sensing, stress response, and cell growth. However, its contribution to cardiac hypertrophic growth and heart failure remains incompletely understood. Here, we show that the HBP is induced in cardiomyocytes during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service