Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0001634M1

Sigma-Aldrich

Monoclonal Anti-DCN antibody produced in mouse

clone 3H4-1F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DSPG2, Anti-PG40, Anti-PGII, Anti-PGS2, Anti-SLRR1B, Anti-decorin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H4-1F4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCN(1634)

General description

Decorin (DCN) protein is encoded by the gene mapped to human chromosome 12q21.33. DCN is an extracellular matrix protein with leucine-rich repeats. The protein belongs to the small leucine-rich proteoglycan (SLRP) gene family.

Immunogen

DCN (AAH05322, 1 a.a. ~ 359 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MKATIILLLLAQVSWAGPFQQRGLFDFMLEDEASGIGPEVPDDRDFEPSLGPVCPFRCQCHLRVVQCSDLGLDKVPKDLPPDTTLLDLQNNKITEIKDGDFKNLKNLHALILVNNKISKVSPGAFTPLVKLERLYLSKNQLKELPEKMPKTLQELRAHENEITKVRKVTFNGLNQMIVIELGTNPLKSSGIENGAFQGMKKLSYIRIADTNITSIPQGLPPSLTELHLDGNKISRVDAASLKGLNNLAKLGLSFNSISAVDNGSLANTPHLRELHLDNNKLTRVPGGLAEHKYIQVVYLHNNNISVVGSSDFCPPGHNTKKASYSGVSLFSNPVQYWEIQPSTFRCVYVRSAIQLGNYK

Biochem/physiol Actions

Decorin (DCN) acts as a structural and signaling molecule and is involved in extracellular matrix assembly. It is implicated in the regulation of fibrogenesis and inflammation. DCN exhibits antimetastatic and antiangiogenic activities by inducing cell cycle arrest and apoptosis. It is used as a marker to detect the prognosis of various types of cancers such as breast, bladder, and lung cancer. Mutation in the DCN gene is associated with the development of posterior amorphous corneal dystrophy (PACD).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madeline Yung et al.
Ophthalmic genetics, 42(4), 486-492 (2021-05-19)
Concomitant corneal ectasia and posterior lamellar corneal opacification is rare, and the genetic relationship between these two conditions is unclear. We report the genetic and clinical characterization of this phenotype in three unrelated individuals. One previously reported affected individual and
Kornélia Baghy et al.
Advances in experimental medicine and biology, 1272, 17-38 (2020-08-28)
The tumor microenvironment plays a determining role in cancer development through a plethora of interactions between the extracellular matrix and tumor cells. Decorin is a prototype member of the SLRP family found in a variety of tissues and is expressed
Wen Zhang et al.
Oncotarget, 9(4), 5480-5491 (2018-02-13)
Decorin (DCN), an extracellular matrix (ECM) protein, belongs to the small leucine-rich proteoglycan family. As a pluripotent molecule, DCN regulates the bioactivities of cell growth factors and participates in ECM assembly. Accumulating evidence has shown that DCN acts as a
Fuqiong Chen et al.
Frontiers in endocrinology, 11, 479258-479258 (2020-12-29)
Cardiomyopathy is the leading cause of increased mortality in diabetes. In the present study, we investigated the effects of decorin (DCN) gene therapy on left ventricular function, cardiac inflammation and fibrosis in type 2 diabetes. Type 2 diabetes was induced
Razie Rezaie et al.
Iranian biomedical journal, 24(4), 229-235 (2020-04-21)
Numerous studies confirmed that significant decrease in tissue decorin (DCN) expression is associated to tumor progression and metastasis in certain types of cancer including prostate cancer (PC). However, the potential prognostic value of tissue DCN in PC has not yet

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service