Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W3000

Sigma-Aldrich

Wright stain

certified by the Biological Stain Commission

Synonym(s):

Eosin Methylene blue according to Wright

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500

grade

certified by the Biological Stain Commission

form

powder

solubility

methanol: 1 mg/mL

InChI

1S/C20H8Br4O5.C16H18N3S/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13/h1-6H,(H2-,25,26,27,28);5-10H,1-4H3/q;+1/p+1

InChI key

AXIKDPDWFVPGOD-UHFFFAOYSA-O

Looking for similar products? Visit Product Comparison Guide

Application

Reported to be superior to May-Grunwald Stain in blood staining.

Suitability

Certified as a stain for blood smears; and as a tissue stain in Nocht′s method. The Wright stain solution is used in place of the separate eosin and azure solutions in the Nocht method.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis and testing of biological stains--the Biological Stain Commission Procedures.
D P Penney et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 77(5-6), 237-275 (2003-02-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service