Skip to Content
MilliporeSigma
All Photos(1)

Documents

VIV403008

SynVivo SynTumour 3D cancer mode starter kit, microvascular network configuration, 8 um pillars, includes pneumatic priming device

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NA.84

packaging

case of 1 ea

manufacturer/tradename

SynVivo 403008

storage temp.

room temp

General description

10 - SMN2 C002 SynTumor 50 μm gap, 8 μm height pillar barrier chips, 100 ft Tygon Tubing, 25 pack - Slide Clamps, 50 pack - 1 mL Syringe with Luer-Lok Tip, 50 pack - 24 Gauge Needle x 0.5 in long, SynVivo Pneumatic Primer, 2 - Five Stopcock Manifold 4-Way all FLL

SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment. The system enables (a) circulation in the microvasculature, (b) transport across the vessel walls, and (c) delivery to the tumors. Starting with scans of vascular networks incorporated with interstitial and tissue/tumor spaces, the SynTumor 3D tissue model creates an in-vitro tumor microenvironment akin to a viable histological slice.

The SynTumor 3D Cancer model has the following benefits:

  • Side by side architecture enables quantitative real time visualization
  • Physiological leaky vasculature with engineered porous structures
  • Morphologically realistic in vivo based architecture
  • Physiologically realistic convective and diffusive transport
Microfluidic platform with ultra-low consumable volumes

Linkage

LInk to Publications
application text

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service