Skip to Content
MilliporeSigma
All Photos(1)

Documents

VIV401003

SynVivo SynRAM 3D Inflammation model assay kit, microvascular network configuration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NA.84

packaging

case of 1 ea

manufacturer/tradename

SynVivo 401003

storage temp.

room temp

General description

10 - SMN2 C001 SynRAM 50 μm gap, 3 μm height pillar barrier chips, 100 ft Tygon Tubing, 25 pack Slide Clamps, 50 pack 1 mL Syringe with Luer-Lok Tip, 50 pack 24 Gauge Needle x 0.5 in long

The SynRAM 3D Inflammation Model from SynVivo has been developed to study the entire inflammation pathway in a realistic and dynamic environment. By recreating a histological slice of co-cultured tissue and/or tumor cells with a lumen of endothelial cells, the SynVivo platform delivers a physiologically realistic model including flow and shear in a platform and enables real-time tracking of rolling, adhesion and migration processes. This model has been successfully validated against in vivo studies showing excellent correlation with rolling velocities, adhesion patterns and migratory processes (Lamberti et al 2014, Soroush et al 2016).

The SynRAM 3D inflammation model provides a realistic testing environment including:

  • Physiological shear stress within a microvascular environment
  • In vivo like vascular morphology with fully enclosed lumen
  • Co-culture capability for cell-cell interactions
  • Quantitative real-time rolling, adhesion, and migration data from a single experiment

SynRAM′s innovative design overcomes the current limitations inherent in flow chambers or Transwell chamber based assays. Current flow chamber designs are oversimplified, lack the scale and geometry of the microenvironment and cannot model transmigration. Similarly, Transwell chambers do not account for fluid shear and size/topology observed in vivo, the end point measurements of migration are not reproducible and do not provide real-time visualization.

SynVivo′s proprietary chip designs range from complex in vivo derived microvascular networks (obtained from digitized images) to produce realistic cellular makeup and vascular morphology resulting in varying shear and flow conditions, to simplified idealized networks designed to reproduce the cellular makeup and constant shear and flow conditions.

Linkage

LInk to Publications
application text

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service