Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V5627

Sigma-Aldrich

Val-Ser

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CC(C)C(N)C(=O)NC(CO)C(O)=O

InChI

1S/C8H16N2O4/c1-4(2)6(9)7(12)10-5(3-11)8(13)14/h4-6,11H,3,9H2,1-2H3,(H,10,12)(H,13,14)

InChI key

STTYIMSDIYISRG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A non-planar peptide bond in L-seryl-L-valine.
Anders Moen et al.
Acta crystallographica. Section C, Crystal structure communications, 60(Pt 8), o564-o565 (2004-08-06)
L-valyl-L-serine trihydrate.
Johansen A, Midtkandal R, Roggen H, Gorbitz CH.
Acta Crystallographica Section C, Crystal Structure Communications, 61, 198-200 (2005)
Studies on the conformation of amino acids. IX. Conformations of butyl, seryl, threonyl, cystennyl, and valyl residues in a dipeptide unit.
P K Ponnuswamy et al.
Biopolymers, 10(3), 565-582 (1971-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service