Skip to Content
MilliporeSigma
All Photos(1)

Documents

V2131

Sigma-Aldrich

Val-Arg-Lys-Arg-Thr-Leu-Arg-Arg-Leu

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H100N22O11
CAS Number:
Molecular Weight:
1197.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

composition

Peptide content, ≥65%

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(C)C)NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCCN)NC(=O)C(CCCNC(N)=N)NC(=O)C(N)C(C)C)C(C)O)C(O)=O

Substrates

Protein kinase C substrate

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C House et al.
The Journal of biological chemistry, 262(2), 772-777 (1987-01-15)
The substrate specificity of protein kinase C has been examined using a series of synthetic peptide analogs of glycogen synthase, ribosomal protein S6, and the epidermal growth factor receptor. The glycogen synthase analog peptide Pro1-Leu-Ser-Arg-Thr-Leu-Ser-Val-Ala-Ala10 was phosphorylated at Ser7 with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service