Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V0256

Sigma-Aldrich

Val-Pro-Asp-Pro-Arg

≥97% (HPLC)

Synonym(s):

Enterostatin (pig, rat)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H42N8O8
CAS Number:
Molecular Weight:
582.65
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

storage temp.

−20°C

Biochem/physiol Actions

An appetite suppressant in rats

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Erlanson-Albertsson et al.
Regulatory peptides, 22(4), 325-331 (1988-09-01)
Pancreatic procolipase is a cofactor for lipase and necessary for optimal fat digestion in the intestine during a meal. It is activated by trypsin in the intestine during release of an activation peptide, with the sequence Val-Pro-Asp-Pro-Arg in rat. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service