Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

UC184

Sigma-Aldrich

Progesterone 11α-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38O9
Molecular Weight:
506.59
UNSPSC Code:
12161501
NACRES:
NA.77

form

solid

color

off-white

mp

208-218 °C

solubility

methanol: soluble

storage temp.

2-8°C

Biochem/physiol Actions

Phase II metabolite of progesterone

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Progesterone 11α-glucuronide is soluble in methanol.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service