Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

UC178

Sigma-Aldrich

(±)-Nirvanol

Synonym(s):

(±)-5-Ethyl-5-phenyl-2,4-imidazolidinedione, (±)-5-Ethyl-5-phenylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:

form

solid

Quality Level

color

off-white

solubility

acetone: soluble
ethanol: soluble
methanol: soluble

storage temp.

2-8°C

SMILES string

CCC1(NC(=O)NC1=O)c2ccccc2

InChI

1S/C11H12N2O2/c1-2-11(8-6-4-3-5-7-8)9(14)12-10(15)13-11/h3-7H,2H2,1H3,(H2,12,13,14,15)

InChI key

UDTWZFJEMMUFLC-UHFFFAOYSA-N

Biochem/physiol Actions

CYP2B6 N-demethyl metabolite of (±)-mephenytoin; anticonvulsive; hypnotic

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

(±)-Nirvanol is soluble in methanol, acetone and ethanol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B F Bourgeois et al.
Epilepsia, 27(4), 412-418 (1986-07-01)
Stereoselective metabolism has been demonstrated for mephenytoin (MHT), R-MHT being demethylated to the pharmacologically active metabolite 5-phenyl-5-ethylhydantoin (PEH; nirvanol), and S-MHT undergoing aromatic hydroxylation to 4-OH-MHT, with formation of an intermediate arene oxide metabolite. PEH is responsible for the therapeutic
A Pezeshk et al.
Journal of inorganic biochemistry, 42(4), 267-272 (1991-06-01)
The preparation and spectral properties of copper(II) complexes of two hydantoins are reported. Complexes of the general formula Cu(hyd)2(py)2, where hyd = phenytoin or nirvanol; and py = pyridine were prepared and characterized by infrared and ESR. Spectral data show
H Fujioka et al.
Journal of pharmacobio-dynamics, 9(3), 303-314 (1986-03-01)
Disposition of 1-benzenesulfonyl-5,5-diphenylhydantoin (II) having a potent anti-inflammatory activity was compared with that of 5,5-diphenylhydantoin (I), an antiepileptic drug, in order to elucidate whether the pharmacodynamic difference between them can be explained by their physicochemical and pharmacokinetic properties. After oral
W Kalow
Xenobiotica; the fate of foreign compounds in biological systems, 16(5), 379-389 (1986-05-01)
The antiepileptic drug mephenytoin is a racemate. Mephenytoin hydroxylation is a stereospecific reaction and is confined to the S-enantiomer, which is normally eliminated within hours, allowing the R-enantiomer to accumulate since it can be eliminated only within days or weeks.
W H Theodore et al.
Neurology, 34(8), 1100-1102 (1984-08-01)
We investigated the conversion of mephenytoin to nirvanol in five patients with uncontrolled complex partial seizures. After a 50-mg single oral dose, mean peak mephenytoin level was 0.48 microgram/ml and nirvanol 0.37 microgram/ml. After 400 mg, peak mephenytoin level was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service