Documents
TMO382019-0125
Nalgene® Certified Clean bottles
clear PETG, volume 125 mL, case of 48 ea
Synonym(s):
PETG bottle, certified nalgene bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
clear PETG
packaging
case of 48 ea
manufacturer/tradename
Nalgene 382019-0125
capacity
125 mL
volume
125 mL
General description
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries