Documents
TMO3110-0500
Nalgene® high-speed centrifuge tubes
round bottom natural polypropylene copolymer, volume 50 mL, without cap, non-sterile, case of 100 ea
Synonym(s):
Nalgene tubes, round bottom tubes
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
round bottom natural polypropylene copolymer
sterility
non-sterile
feature
without cap
packaging
case of 100 ea
manufacturer/tradename
Nalgene 3110-0500
volume
50 mL
General description
Improve rotor fit and performance using Thermo Scientific Nalgene High-Speed Round-Bottom PPCO Centrifuge Tubes, featuring uniform wall thickness and diameter. Tubes have excellent chemical resistance.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries