Key Documents
TMO2116-0250
Nalgene® straight-sided wide-mouth jar
polycarbonate, jar capacity 250 mL, clear, case of 24 ea
Synonym(s):
clear plastic container, screw top container
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
clear
polycarbonate
straight
wide-mouth
packaging
case of 24 ea
manufacturer/tradename
Nalge-Nunc 2116-0250
jar capacity
250 mL
General description
Collect and store pastes, solids, specimens and lab supplies with window-clear Thermo Scientific Nalgene Straight-side Wide-Mouth Jars. A wide temperature range (-135°C to +135°C) makes these jars an excellent choice for cold room and refrigerated use all the way to the autoclave. These high impact-resistant jars are extremely versatile. Not liquid leakproof.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)