Key Documents
TMO1201-4000
Nalgene® Griffin beakers
polypropylene, volume 4000 mL, case of 4 ea
Synonym(s):
PP beaker, nalgene PP beaker
Select a Size
4 EACH
$445.00
$445.00
Please contact Customer Service for Availability
Select a Size
Change View
4 EACH
$445.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$445.00
Please contact Customer Service for Availability
material
polypropylene
packaging
case of 4 ea
manufacturer/tradename
Nalgene 1201-4000
volume
4000 mL
General description
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Other Notes
Browse all plastic beakers
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)