Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T9632

Sigma-Aldrich

Trizma® succinate

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol succinate, Di(tris[hydroxymethyl]aminomethane) succinate, Tris succinate, Tris(hydroxymethyl)aminomethane succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2C(CH2OH)3]2 · C4H6O4
CAS Number:
Molecular Weight:
360.36
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

Assay

≥98% (titration)

form

crystalline powder

solubility

water: 1 g/mL, clear, colorless

SMILES string

NC(CO)(CO)CO.NC(CO)(CO)CO.OC(=O)CCC(O)=O

InChI

1S/2C4H11NO3.C4H6O4/c2*5-4(1-6,2-7)3-8;5-3(6)1-2-4(7)8/h2*6-8H,1-3,5H2;1-2H2,(H,5,6)(H,7,8)

InChI key

CFJZQNZZGQDONE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service