Skip to Content
MilliporeSigma
All Photos(1)

Documents

T9011

Sigma-Aldrich

Trypsinogen, PMSF treated from bovine pancreas

For use as a marker in SDS-PAGE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

bovine pancreas

Quality Level

form

powder

mol wt

~24 kDa

packaging

vial of 25 mg

application(s)

detection

storage temp.

2-8°C

Related Categories

Application

Trypsinogen, PMSF treated from bovine pancreas has been used as a non-phosphorylated control to analyze the phospho-protein α-casein from bovine milk by sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE). It has also been used as a molecular weight marker standard in SDS-PAGE.

Biochem/physiol Actions

Trypsinogen is a pancreatic proteinase and exists as isoenzymes 1 and 2. It is an inactive precursor of trypsin and gets activated by the enteropeptidase found on the brush border membrane of enterocytes in the duodenum. It is also known to proteolytically activate itself. Trypsinogen activation is associated with acute pancreatitis as it causes pancreatic enzyme cascade which activates the remaining zymogens leading to autodigestion of the neighboring tissue. Hereditary pancreatitis was shown to be caused by a Arg-His substitution at residue 117 of trypsinogen causing auto-activation of trypsinogen to trypsin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D C Whitcomb et al.
Nature genetics, 14(2), 141-145 (1996-10-01)
Hereditary pancreatitis (HP) is a rare, early-onset genetic disorder characterized by epigastric pain and often more serious complications. We now report that an Arg-His substitution at residue 117 of the cationic trypsinogen gene is associated with the HP phenotype. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service