Skip to Content
MilliporeSigma
All Photos(1)

Documents

T8444

Sigma-Aldrich

Anti-phospho-Tau (pSer199) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 60 kDa

species reactivity

human, mouse, rat

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer199)

Gene Information

human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)

Immunogen

chemically synthesized phosphopeptide derived from the region of human tau that contains serine 199.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 50% glycerol, 1.0 mg/ml BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kutay Deniz Atabay et al.
Journal of neurochemistry, 120(3), 430-439 (2011-03-30)
Abnormal cell cycle events are increasingly becoming important attributes of neurodegenerative pathology. Pin1 is a crucial target of neurodegeneration in relation to its functions regarding these abnormal cell cycle events in neurons. Pin1 is majorly involved in many aspects of
Sirin Korulu et al.
The European journal of neuroscience, 37(10), 1610-1619 (2013-03-16)
Neurons are differentiated postmitotic cells residing in G0 phase of the cell cycle and are unable to proceed through G1 phase, in which cyclinD1 needs to be up-regulated for initiation. Yet, a growing body of evidence has shown that cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service