Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T8132

Sigma-Aldrich

2,2,2-Trifluoroethanol

Synonym(s):

Trifluoroethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2OH
CAS Number:
Molecular Weight:
100.04
Beilstein:
1733203
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

3.5 (vs air)

vapor pressure

70 mmHg ( 25 °C)

Assay

≥99%

autoignition temp.

~896 °F

expl. lim.

42 %

refractive index

n20/D 1.3 (lit.)

bp

77-80 °C (lit.)

mp

−44 °C (lit.)

density

1.373 g/mL at 25 °C (lit.)

SMILES string

OCC(F)(F)F

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2

InChI key

RHQDFWAXVIIEBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service