Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7783

Sigma-Aldrich

Thonzonium bromide

Synonym(s):

Hexadecyl(2-[(p-methoxybenzyl)-2-pyrimidinylamino]ethyl)dimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C32H55N4OBr
CAS Number:
Molecular Weight:
591.71
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

cationic

mol wt

591.71 g/mol

application(s)

sample preparation

SMILES string

Br.CCCCCCCCCCCCCCCC[N](C)(C)CCN(Cc1ccc(OC)cc1)c2ncccn2

InChI

1S/C32H55N4O.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-27-36(2,3)28-26-35(32-33-24-19-25-34-32)29-30-20-22-31(37-4)23-21-30;/h19-25H,5-18,26-29H2,1-4H3;1H

InChI key

PBWQBSCBSFJTLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thonzonium bromide may be use to inhibit vacuolar ATPases (V-ATPases) to induce and study the effects of cytosolic acidification. Thonzonium bromide is also used to further characterize V-ATPases. Thonzonium bromide is use as a potential pDNA particle condenser.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A multifunctional nano device as non-viral vector for gene delivery: in vitro characteristics and transfection.
Gao Y, Gu W, Chen L, Xu Z, Li Y.
J. Controlled Release, 118, 381-388 (2007)
Ryosuke Suzuki et al.
Biological & pharmaceutical bulletin, 31(6), 1237-1243 (2008-06-04)
Non-viral DNA vectors are promising gene delivery systems and a variety of non-viral DNA vectors have been developed to date. Recently, we developed a novel non-viral gene delivery system--multifunctional envelope-type nano device (MEND). The MEND system has high transfection activity
Chun-Yuan Chan et al.
The Journal of biological chemistry, 287(13), 10236-10250 (2012-01-05)
Vacuolar ATPases (V-ATPases) are important for many cellular processes, as they regulate pH by pumping cytosolic protons into intracellular organelles. The cytoplasm is acidified when V-ATPase is inhibited; thus we conducted a high-throughput screen of a chemical library to search
Rebecca M Johnson et al.
Analytical biochemistry, 398(2), 203-211 (2009-12-19)
Fluorescence intensity of the pH-sensitive carboxyfluorescein derivative 2,7-bis(2-carboxyethyl)-5(6)-carboxyfluorescein (BCECF) was monitored by high-throughput flow cytometry in living yeast cells. We measured fluorescence intensity of BCECF trapped in yeast vacuoles, acidic compartments equivalent to lysosomes where vacuolar proton-translocating ATPases (V-ATPases) are
Cristiana Lalli et al.
PLoS neglected tropical diseases, 9(1), e0003484-e0003484 (2015-01-31)
Schistosomiasis, one of the world's greatest neglected tropical diseases, is responsible for over 280,000 human deaths per annum. Praziquantel, developed in the 1970s, has high efficacy, excellent tolerability, few and transient side effects, simple administration procedures and competitive cost and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service