Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7425

Sigma-Aldrich

DL-Tryptophan

suitable for cell culture, ≥99% (TLC)

Synonym(s):

(±)-α-Amino-3-indolepropionic acid, (±)-2-Amino-3-(3-indolyl)propionic acid, DL-3β-Indolylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Beilstein:
86199
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥99% (TLC)

technique(s)

cell culture | mammalian: suitable

mp

289-290 °C (dec.) (lit.)

SMILES string

NC(Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)

InChI key

QIVBCDIJIAJPQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in insect and mammalian cell culture applications where the pure L-form is not required.

Preparation Note

Prepared from L-tryptophan produced by fermentation.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yekta Dowlati et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 25(6), 779-787 (2015-04-01)
Postpartum depression (PPD) is the most common complication of childbearing with a 13% prevalence rate. Sleep disturbances are also common, particularly during early postpartum. In theory, l-tryptophan could improve sleep and reduce depressed mood in early postpartum; however, the first
Helen Ling et al.
Movement disorders : official journal of the Movement Disorder Society, 30(7), 960-967 (2015-04-10)
Glial cytoplasmic inclusions containing α-synuclein are the pathological hallmark of multiple system atrophy (MSA). Minimal change (MC-MSA) is an unusual MSA subtype with neuronal loss largely restricted to the substantia nigra and locus coeruleus. Immunohistochemistry on selected brain regions and
Aristeidis Chiotellis et al.
Molecular pharmaceutics, 11(11), 3839-3851 (2014-07-06)
As a continuation of our research efforts toward the development of tryptophan-based radiotracers for tumor imaging with positron emission tomography (PET), three new fluoroethoxy tryptophan analogues were synthesized and evaluated in vivo. These new tracers (namely, 4-(2-[(18)F]fluoroethoxy)-dl-tryptophan ([(18)F]4-FEHTP), 6-(2-[(18)F]fluoroethoxy)-dl-tryptophan ([(18)F]6-FEHTP)
Kazuo Tatebayashi et al.
Nature communications, 6, 6975-6975 (2015-04-22)
The yeast high osmolarity glycerol (HOG) pathway activates the Hog1 MAP kinase, which coordinates adaptation to high osmolarity conditions. Here we demonstrate that the four-transmembrane (TM) domain protein Sho1 is an osmosensor in the HKR1 sub-branch of the HOG pathway.
C C K Hui et al.
Mucosal immunology, 8(5), 993-999 (2014-12-18)
Recent candidate gene and genome-wide association studies have identified "protective" associations between the single-nucleotide polymorphism (SNP) rs1837253 in the TSLP gene and risk for allergy, asthma, and airway hyperresponsiveness. The absence of linkage disequilibrium of rs1837253 with other SNPs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service