Skip to Content
MilliporeSigma
All Photos(1)

Documents

T7076

Sigma-Aldrich

Monoclonal Anti-Thimet Oligopeptidase 1 antibody produced in mouse

1 mg/mL, clone IVD6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-THOP1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IVD6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~85 kDa

species reactivity

human, monkey

concentration

1 mg/mL

technique(s)

microarray: suitable
western blot: 0.5-1.0 μg/mL using SY5Y cell extract.

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... THOP1(7064)

General description

Monoclonal Anti-Thimet Oligopeptidase 1 (THOP1) (mouse IgG2b isotype) is derived from the hybridoma IVD6 produced by the fusion of mouse myeloma cells (SP2/O cells) and splenocytes from CF1mice immunized with a synthetic peptide. THOP1 protein is found in tissues with high levels of neuropeptides and hormones. THOP1 gene is mapped on the human chromosome at 19p13.3.

Specificity

Monoclonal Anti-Thimet Oligopeptidase recognizes human and monkey THOP1, ~85 kDa.

Immunogen

synthetic peptide corresponding to amino acids 47-63 of human THOP1.

Application

Monoclonal Anti-Thimet Oligopeptidase 1 antibody produced in mouse may be used in:
  • immunoblotting
  • immunocytochemistry
  • immunohistochemistry
  • enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Thimet Oligopeptidase 1 (THOP1) is a metalloendopeptidase found in the brains of Alzheimer patients. It can cleave a synthetic peptide encompassing the amino-terminus of amyloid β-peptide and generate amyloidogenic fragments from recombinant human amyloid precursor protein (APP). THOP1 is involved in degrading peptides that are released from the proteasome and therefore limits the extent of major histocompatibility complex (MHC) class I antigen presentation. It is involved in the processing of bioactive peptides. Overexpression of the gene THOP1 in cells reduced the presentation of antigenic peptides by class I MHC molecules. Prevention of THOP1 expression by small interfering RNA promoted class I antigenic peptide presentation.

Target description

THOP1 is a metalloprotease that is able to cleave a synthetic peptide encompassing the amino-terminus of amyloid b-peptideand generate amyloidogenic fragments from recombinant human Amyloid Precursor Protein.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The cytosolic endopeptidase, thimet oligopeptidase, destroys antigenic peptides and limits the extent of MHC class I antigen presentation
York I A, et al.
Immunity, 18(3), 429-440 (2003)
Comparative maps of human 19p13. 3 and mouse chromosome 10 allow identification of sequences at evolutionary breakpoints
Puttagunta R, et al.
Genome Research, 10(9), 1369-1380 (2000)
Giuseppe Pollio et al.
Neurobiology of disease, 31(1), 145-158 (2008-06-24)
In a comprehensive proteomics study aiming at the identification of proteins associated with amyloid-beta (Abeta)-mediated toxicity in cultured cortical neurons, we have identified Thimet oligopeptidase (THOP1). Functional modulation of THOP1 levels in primary cortical neurons demonstrated that its overexpression was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service