Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7012

Sigma-Aldrich

Tetraethylammonium bromide

≥99%

Synonym(s):

TEA bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Br)
CAS Number:
Molecular Weight:
210.16
Beilstein:
3563430
EC Number:
MDL number:
UNSPSC Code:
12161900

Assay

≥99%

mp

285 °C (dec.) (lit.)

SMILES string

[Br-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

HWCKGOZZJDHMNC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service