Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

T6507

Sigma-Aldrich

Tridecyl acetate

liquid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30O2
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

form

liquid

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCOC(C)=O

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Guo et al.
Journal of insect science (Online), 9, 1-9 (2010-01-08)
Olfactory perception of myristic, palmitic, stearic and oleic acids and their corresponding methyl esters by Asian corn borer moths, Ostrinia furnacalis (Guenée) (Lepidoptera: Crambidae) was investigated. It was found that mated females with both antennae amputated, in contrast to intact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service