Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6269

Sigma-Aldrich

o-Tolidine dihydrochloride

≥97%

Synonym(s):

3,3′-Dimethylbenzidine dihydrochloride, 4,4′-Bianisidine dihydrochloride, 4,4′-Diamino-3,3′-dimethylbiphenyl dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H3(CH3)-4-NH2]2 · 2HCl
CAS Number:
Molecular Weight:
285.21
Beilstein:
3713238
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97%

color

white to light yellow

SMILES string

Cl.Cl.Cc1cc(ccc1N)-c2ccc(N)c(C)c2

InChI

1S/C14H16N2.2ClH/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12;;/h3-8H,15-16H2,1-2H3;2*1H

InChI key

LUKPNZHXJRJBAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sensitive colorimetric reagent for gold and for free chlorine in water.

Warning

May be carcinogenic.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service