Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6025

Sigma-Aldrich

Tris Acetate-EDTA buffer

BioReagent, suitable for electrophoresis

Synonym(s):

TAE buffer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.25

sterility

0.2 μm filtered

Quality Level

product line

BioReagent

form

working solution

technique(s)

electrophoresis: suitable

impurities

DNase, RNase, Protease, none detected

suitability

suitable for electrophoresis
suitable for gel electrophoresis (after dilution to working concentration)

application(s)

diagnostic assay manufacturing

SMILES string

CC(O)=O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3.C2H4O2/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8;1-2(3)4/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2;1H3,(H,3,4)

InChI key

HGEVZDLYZYVYHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis. Double-stranded DNA tends to run faster in TAE than in other buffers but can also become exhausted during extended electrophoresis. Buffer circulation or buffer replacement during extended electrophoresis can remedy the lower buffering capacity. Dilution of the concentrated TAE buffer produces a 1× TAE buffer with 40 mM Tris-acetate and 1 mM EDTA, pH 8.3. The 1× TAE buffer is used both in the agarose gel and as a running buffer. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.
Tris Acetate-EDTA buffer has been used as run buffer for the agarose gel electrophoresis of human papillomavirus DNA and transcribed RNA samples.

Other Notes

40 mM Tris acetate, pH approx. 8.3, containing 1 mM EDTA.

Preparation Note

Prepared with Biotechnology Performance Certified Trizma base (T6066) and Molecular Biology Reagent EDTA disodium salt (E5134). Solutions also contain acetic acid (A6283); powdered blend contains Trizma acetate (T1258).
Solution prepared with 18 megohm water

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An in vitro enzymatic assay to measure transcription inhibition by gallium (III) and H3 5, 10, 15-tris (pentafluorophenyl) corroles
Tang GY, et al.
Journal of Visualized Experiments, 2(97), e52355-e52355 (2015)
Monica Musiani et al.
Nature protocols, 2(10), 2502-2510 (2007-10-20)
PCR is an established technique providing rapid and highly productive amplification of specific DNA sequences. The demand for equally rapid, sensitive and objective methods to achieve detection of PCR products has led to the coupling of PCR with ELISA. PCR-ELISA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service