Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5824

Sigma-Aldrich

TAN-67 dihydrobromide

≥98% (HPLC), solid

Synonym(s):

(3-Hydroxyphenyl)-1,2,3,4,4a,5,12,12aa-octahydroquinolino[2,3,3-g]isoquinoline dihydrobromide, SB-205607 dihydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24N2O · 2HBr
CAS Number:
Molecular Weight:
506.27
MDL number:
UNSPSC Code:
12352200

description

Store tightly sealed under nitrogen

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

H2O: soluble >20 mg/mL

storage temp.

2-8°C

SMILES string

Br[H].Br[H].[H][C@@]12CN(C)CC[C@]1(Cc3nc4ccccc4cc3C2)c5cccc(O)c5

Biochem/physiol Actions

Potent, selective non-peptide δ opioid receptor agonist.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemal H Patel et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 16(11), 1468-1470 (2002-09-03)
Opioids and volatile anesthetics produce marked cardioprotective effects against myocardial infarction via the activation of ATP-sensitive potassium (K(ATP)) channels, however, the effect of combined treatment with both drugs is unknown. We examined the hypothesis that opioids and volatile anesthetics potentiate
R M Fryer et al.
The Journal of pharmacology and experimental therapeutics, 299(2), 477-482 (2001-10-17)
We investigated the possibility that opioids activate a tyrosine kinase (TK) that mediates cardioprotection in an in vivo rat model of myocardial infarction. All animals underwent 30 min of regional ischemia and 2 h of reperfusion. Infarct size was expressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service