Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5780

Sigma-Aldrich

DL-Thyronine

Synonym(s):

3-(p-[p-Hydroxyphenoxy]phenyl)-DL-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO4
CAS Number:
Molecular Weight:
273.28
EC Number:
MDL number:
UNSPSC Code:
12352200

SMILES string

NC(Cc1ccc(Oc2ccc(O)cc2)cc1)C(O)=O

InChI

1S/C15H15NO4/c16-14(15(18)19)9-10-1-5-12(6-2-10)20-13-7-3-11(17)4-8-13/h1-8,14,17H,9,16H2,(H,18,19)

InChI key

KKCIOUWDFWQUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Similar to T 1501, but produced by Sigma.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service