Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5568

Sigma-Aldrich

Tyrphostin AG 835

≥92%

Synonym(s):

(+)-(S)-N-(α-Methylbenzyl)-3,4-dihydroxybenzylidenecyanoacetamide, Tyrphostin B50

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O3
CAS Number:
Molecular Weight:
308.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥92%

form

crystalline

potency

IC50 (860 nanomolar for the inhibition of EGFR-kinase activity.)

solubility

DMSO or ethanol: soluble

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)\C(=C\c1ccc(O)c(O)c1)C#N)c2ccccc2

InChI

1S/C18H16N2O3/c1-12(14-5-3-2-4-6-14)20-18(23)15(11-19)9-13-7-8-16(21)17(22)10-13/h2-10,12,21-22H,1H3,(H,20,23)/b15-9+/t12-/m0/s1

InChI key

UMGQVUWXNOJOSJ-DGGAMASNSA-N

Biochem/physiol Actions

Protein tyrosine kinase inhibitor.

Preparation Note

Tyrphostin AG 835 is soluble in ethanol and DMSO.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service