Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

T5441

Sigma-Aldrich

TAPS sodium salt

BioPerformance Certified, suitable for cell culture, ≥99%

Synonym(s):

N-tris(Hydroxymethyl)methyl-3-aminopropanesulfonic acid sodium-potassium salt, [(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid sodium-potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16NNaO6S
CAS Number:
Molecular Weight:
265.26
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified
Molecular Biology
for molecular biology

Assay

≥99%

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and bioburden, tested
≤1% water

useful pH range

7.7-9.1

pKa (25 °C)

8.4

solubility

water: 0.33 g/mL, clear to slightly hazy, colorless

cation traces

heavy metals (as Pb): ≤5 ppm

application(s)

diagnostic assay manufacturing

foreign activity

DNase, RNase, protease, none detected

SMILES string

[Na+].OCC(CO)(CO)NCCCS([O-])(=O)=O

InChI

1S/C7H17NO6S.Na/c9-4-7(5-10,6-11)8-2-1-3-15(12,13)14;/h8-11H,1-6H2,(H,12,13,14);/q;+1/p-1

InChI key

FEGYIWVHCSRXCG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service