Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5153

Sigma-Aldrich

Tyr-Gly-Ala-Val-Val-Asn-Asp-Leu

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H59N9O13
CAS Number:
Molecular Weight:
849.93
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B M Dutia et al.
Nature, 321(6068), 439-441 (1986-05-22)
Ribonucleotide reductase is an essential enzyme for DNA synthesis in all prokaryotic and eukaryotic cells; it catalyses the reductive conversion of ribonucleotides to deoxyribonucleotides. Several herpesviruses including herpes simplex virus type 1 (HSV-1), HSV-2, pseudorabies virus (PRV), equine herpesvirus type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service