Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5007

Sigma-Aldrich

Glyceryl trilinolelaidate

~98% (capillary GC), liquid

Synonym(s):

1,2,3-Tri-([trans,trans]-9,12-octadecadienoyl)glycerol, Trilinolelaidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H98O6
CAS Number:
Molecular Weight:
879.38
MDL number:
UNSPSC Code:
12352200

Assay

~98% (capillary GC)

form

liquid

storage temp.

−20°C

SMILES string

CCCCC\C=C\C\C=C\CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C\C\C=C\CCCCC)OC(=O)CCCCCCC\C=C\C\C=C\CCCCC

Packaging

Sealed ampule.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ju-Chi Liu et al.
Journal of biomedical science, 11(1), 11-18 (2004-01-20)
The myocardial protective effects of trilinolein, isolated from the traditional Chinese herb Sanchi (Panax notoginseng), are thought to be related to its antioxidant activity. However, the intracellular mechanism underlying the protective effect of trilinolein in the heart remains unclear. In
Paul Chan et al.
Acta pharmacologica Sinica, 23(12), 1157-1162 (2002-12-06)
Trilinolein is a triacylglycerol purified from a commonly used traditional Chinese medicine Panax notoginseng. Trilinolein has been reported to provide a number of beneficial effects including reducing thrombogenicity and arrhythmias and increasing erythrocyte deformability. Additionally, trilinolein has been reported to
Clara De Pascale et al.
The FEBS journal, 276(19), 5689-5702 (2009-09-04)
Current evidence indicates that chylomicron remnants (CMR) induce macrophage foam cell formation, an early event in atherosclerosis. Inflammation also plays a part in atherogenesis and the transcription factor nuclear factor-kappaB (NF-kappaB) has been implicated. In this study, the influence of
Gloria Márquez-Ruiz et al.
Lipids, 38(3), 233-240 (2003-06-06)
The effects of temperature and addition of alpha-tocopherol were evaluated in trilinolein model systems through quantification of oxidized TAG monomers, dimers, and polymers following oxidation at different temperatures. Samples of trilinolein without and with 250 and 500 mg/kg alpha-tocopherol added
Yong Ihl Park et al.
Journal of insect physiology, 50(6), 539-546 (2004-06-09)
After swarming, reproductive dealates of the Formosan subterranean termite, Coptotermes formosanus, run together in tandem. The tandem running is an age-related behavioral activity in C. formosanus. This behavior was not evoked in pre-flight alates less than 6-d-old that were artificially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service