Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T4192

Sigma-Aldrich

Tyrphostin SU 1498

≥98%

Synonym(s):

(E)-N-(3″-Phenylpropyl)-α-cyano-3′,5′-diisopropyl-4′-hydroxycinnamamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N2O2
CAS Number:
Molecular Weight:
390.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

form

solid

potency

700 nM IC50

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

CC(C)c1cc(\C=C(/C#N)C(=O)NCCCc2ccccc2)cc(C(C)C)c1O

InChI

1S/C25H30N2O2/c1-17(2)22-14-20(15-23(18(3)4)24(22)28)13-21(16-26)25(29)27-12-8-11-19-9-6-5-7-10-19/h5-7,9-10,13-15,17-18,28H,8,11-12H2,1-4H3,(H,27,29)/b21-13+

InChI key

JANPYFTYAGTSIN-FYJGNVAPSA-N

Application

Tyrphostin SU 1498 has been used as a VEGF inhibitor in mice1.

Biochem/physiol Actions

Potent and selective inhibitor of the VEGF receptor kinase, Flk-1. Very weak inhibitor of PDGFR-kinase, EGFR-kinase and HER-2 kinase.

Preparation Note

Tyrphostin SU 1498 is soluble in DMSO.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaorui Shi
The American journal of pathology, 174(5), 1692-1704 (2009-04-08)
This study explored the effect of acoustic trauma on cochlear pericytes. Transmission electron microscopy revealed that pericytes on capillaries of the stria vascularis were closely associated with the endothelium in both control guinea pigs and mice. Pericyte foot processes were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service