Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3691

Sigma-Aldrich

Clear-view Snap-Cap microtubes

size 1.5 mL, assorted colors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702

material

assorted colors cap
assorted colors tube
cap (Angled cap lid)
conical bottom
flat cap
polypropylene
tube (homopolymer)

sterility

non-sterile

feature

RCF 20,000 × g
RNase and DNase free
neck thread Push fit

packaging

pack of 500 ea

manufacturer/tradename

Excel Scientific MCT-1.5A

technique(s)

PCR: suitable

capacity

1.5 mL

size

1.5 mL

color

assorted colors

Looking for similar products? Visit Product Comparison Guide

General description

  • Superior clarity for sample viewing
  • A consistent, ring-lock seal prevents inadvertent opening
  • Angled cap lid makes opening easy without thumb fatigue
  • Large flat tops for ease of identification and a pierceable cap
  • Large, crisp reference lines and graduation numbers
  • Certified RNase/DNase free

Legal Information

Clear-view is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin E Mulkearns-Hubert et al.
Cell reports, 27(4), 1062-1072 (2019-04-25)
Gap-junction-mediated cell-cell communication enables tumor cells to synchronize complex processes. We previously found that glioblastoma cancer stem cells (CSCs) express higher levels of the gap junction protein Cx46 compared to non-stem tumor cells (non-CSCs) and that this was necessary and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service