Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3275

Sigma-Aldrich

Thr-Gln

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17N3O5
CAS Number:
Molecular Weight:
247.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CC(O)C(N)C(=O)NC(CCC(N)=O)C(O)=O

InChI

1S/C9H17N3O5/c1-4(13)7(11)8(15)12-5(9(16)17)2-3-6(10)14/h4-5,7,13H,2-3,11H2,1H3,(H2,10,14)(H,12,15)(H,16,17)

InChI key

BWUHENPAEMNGQJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Roturier et al.
Journal of chromatography. A, 696(2), 209-217 (1995-04-14)
Small hydrophilic di- and tripeptides from food products are not separated by reversed-phase high-performance liquid chromatography (RP-HPLC). A simple method using precolumn derivatization with 9-fluorenylmethoxycarbonyl chloride (FMOC) of hydrophilic peptides followed by RP-HPLC separation is presented. Peptides can subsequently be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service