Skip to Content
MilliporeSigma
All Photos(1)

Documents

T2752

Sigma-Aldrich

3,3′,5-Triiodo-L-thyronine sodium salt

≥95% (HPLC)

Synonym(s):

Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11I3NNaO4
CAS Number:
Molecular Weight:
672.96
Beilstein:
8179867
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Assay

≥95% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

ethanol/1 M HCl (4:1): 10 mg/mL, clear, colorless to faintly yellow or pink

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C([O-])=O

InChI

1S/C15H12I3NO4.Na/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22;/h1-4,6,12,20H,5,19H2,(H,21,22);/q;+1/p-1/t12-;/m0./s1

InChI key

SBXXSUDPJJJJLC-YDALLXLXSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

3,3′,5-Triiodo-L-thyronine sodium salt was used in Oligodendrocyte differentiation medium used for isolation and culture of rat and mouse oligodendrocyte precursor cells. The product was used as a test compound for incubating adipocyte cells, used for studying the impact of hormones and butyrate on modulation of porcine adipocyte beta-adrenergic receptors.

Biochem/physiol Actions

3,3′,5-Triiodo-L-thyronine also referred to as O-(4-Hydroxy-3-iodophenyl)−3,5−diiodo-L-tyrosine sodium salt or T3 is the biologically active thyroid hormone. In hypothyroidism pateints, triiodothyronine can enhance mood and neuropsychological function. The product also positively regulates the hepatic FGF21 expression and stimulates the metabolic breakdown of glucose, fats, and proteins by enhancing the levels of numerous metabolic enzymes such as hexokinase, liver glucose 6-phosphatase as well as mitochondrial enzymes for oxidative phosphorylation.

Preparation Note

3,3′,5-Triiodo-L-thyronine sodium salt dissolves in 1 M HCl:EtOH with a ratio of 1:4 at 10 mg/ml to yield a clear, colorless to yellow or faint pink solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Ding et al.
Journal of animal science, 78(4), 927-933 (2000-04-28)
The beta-adrenergic receptors (betaAR) on the surface of mammalian cells are desensitized when the cell is stimulated by betaAR agonists to eliminate excessive response by the cell. Investigations with adipocytes, primarily rodent-derived cells, indicate other hormones and substrates also can
Ye-Seul Yoon et al.
Biomolecules & therapeutics, 29(1), 83-89 (2020-06-17)
Multiple system atrophy (MSA) is a neurodegenerative disease characterized by presence of α-synuclein-positive inclusions in the cytoplasm of oligodendrocytes. These glial cytoplasmic inclusions (GCIs) are considered an integral part of the pathogenesis of MSA, leading to demyelination and neuronal demise.
Andrew C Adams et al.
The Journal of biological chemistry, 285(19), 14078-14082 (2010-03-20)
Thyroid hormone has profound and diverse effects on liver metabolism. Here we show that tri-iodothyronine (T3) treatment in mice acutely and specifically induces hepatic expression of the metabolic regulator fibroblast growth factor 21 (FGF21). Mice treated with T3 showed a
R Bunevicius et al.
The New England journal of medicine, 340(6), 424-429 (1999-02-11)
Patients with hypothyroidism are usually treated with thyroxine (levothyroxine) only, although both thyroxine and triiodothyronine are secreted by the normal thyroid gland. Whether thyroid secretion of triiodothyronine is physiologically important is unknown. We compared the effects of thyroxine alone with
Radim Vrzal et al.
Toxicology letters, 275, 77-82 (2017-05-10)
Aryl hydrocarbon receptor (AhR) is a transcription factor, the activity of which is modulated by hormones including glucocorticoids and estrogens. In this study, we examined the effects of triiodothyronine (T3), a ligand and activator of thyroid hormone receptor (TR), on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service