Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2634

Sigma-Aldrich

β-(2-Thiazolyl)-DL-alanine

≥98% (TLC)

Synonym(s):

(±)-2-Amino-3-(2-thiazolyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2S
CAS Number:
Molecular Weight:
172.20
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

β-(2-Thiazolyl)-DL-alanine,

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

202-204 °C (lit.)

storage temp.

−20°C

SMILES string

NC(Cc1nccs1)C(O)=O

InChI

1S/C6H8N2O2S/c7-4(6(9)10)3-5-8-1-2-11-5/h1-2,4H,3,7H2,(H,9,10)

InChI key

PXFXXRSFSGRBRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Stougaard et al.
Journal of bacteriology, 170(1), 250-257 (1988-01-01)
A histidine auxotrophic (hisA) mutant of Klebsiella pneumoniae is phenotypically Nif- when grown with 20 micrograms of histidine ml-1 but Nif+ when supplied with histidine at 100 micrograms ml-1. Reversion to Nif+ at 20 micrograms of histidine ml-1 occurs phenotypically
F W Pons et al.
Genetics, 120(3), 657-665 (1988-11-01)
Among eight strains carrying acridine-induced mutations in hisH, five which mapped at four different sites in the promoter-distal region of the gene showed His+ phenotypes on media containing a purine. By complementation analysis, hisH enzyme was shown to be required
D E Levin et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(14), 4457-4461 (1984-07-01)
Salmonella tester strain TA102 carries the hisG428 ochre mutation on the multicopy plasmid pAQ1. DNA sequence analysis of 45 spontaneous revertants of hisG428 on the chromosome in the presence of pKM101 (strain TA103) indicates that hisG428 revertants fall into three
J N Picada et al.
Mutation research, 437(2), 165-173 (1999-11-11)
1,2:7,8-Diepoxyoctane (DEO), whose deletogenic activity was first demonstrated in ad-3 system of Neurospora crassa and then in different species, has been tested in Salmonella typhimurium tester strain TA102 (hisG428(Ochre)). It was confirmed that it is a direct acting mutagen and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service