Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1823

Sigma-Aldrich

Tetramethylrosamine chloride salt

≥95% (at 254 nm, HPLC), solid

Synonym(s):

3,6-bis(dimethylamino)-9-phenyl chloride salt, Xanthylium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23N2OCl
Molecular Weight:
378.89
MDL number:
UNSPSC Code:
12161501

description

Light sensitive

Assay

≥95% (at 254 nm, HPLC)

form

solid

solubility

DMSO or DMF: soluble

fluorescence

λem 573 nm (+/- 4nm)

SMILES string

[Cl-].CN(C)c1ccc2c(-c3ccccc3)c4ccc(cc4[o+]c2c1)N(C)C

Application

Useful dye for studying P-gp mediated exclusion from multidrug resistant cells. Superior P-gp substrate compared to the widely used marker rhodamine 123.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service