Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1507

Sigma-Aldrich

Tyr-Tyr-Tyr Methyl ester

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31N3O7
CAS Number:
Molecular Weight:
521.56
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

product name

Tyr-Tyr-Tyr Methyl ester,

Assay

≥98% (TLC)

form

powder

color

white to off-white

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

COC(=O)C(Cc1ccc(O)cc1)NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(N)Cc3ccc(O)cc3

InChI

1S/C28H31N3O7/c1-38-28(37)25(16-19-6-12-22(34)13-7-19)31-27(36)24(15-18-4-10-21(33)11-5-18)30-26(35)23(29)14-17-2-8-20(32)9-3-17/h2-13,23-25,32-34H,14-16,29H2,1H3,(H,30,35)(H,31,36)

InChI key

UXCUYXDJYCOPDL-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service