Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1412

Sigma-Aldrich

Glyceryl tritricosanoate

≥99%

Synonym(s):

1,2,3-Tritricosanoylglycerol, Tritricosanoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H140O6
CAS Number:
Molecular Weight:
1101.88
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99%

form

powder

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C72H140O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-52-55-58-61-64-70(73)76-67-69(78-72(75)66-63-60-57-54-51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)68-77-71(74)65-62-59-56-53-50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h69H,4-68H2,1-3H3

InChI key

JDRVRMABYZMJHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Witting et al.
PloS one, 12(3), e0172311-e0172311 (2017-03-10)
Lipid identification is a major bottleneck in high-throughput lipidomics studies. However, tools for the analysis of lipid tandem MS spectra are rather limited. While the comparison against spectra in reference libraries is one of the preferred methods, these libraries are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service