Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1394

Sigma-Aldrich

Trifluoromethanesulfonic acid

Synonym(s):

Triflic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3H
CAS Number:
Molecular Weight:
150.08
Beilstein:
1812100
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

5.2 (vs air)

vapor pressure

8 mmHg ( 25 °C)

refractive index

n20/D 1.327 (lit.)

bp

162 °C (lit.)

density

1.696 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

OS(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S/c2-1(3,4)8(5,6)7/h(H,5,6,7)

InChI key

ITMCEJHCFYSIIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Deglycosylation agent

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>332.1 °F - Pensky-Martens closed cup

Flash Point(C)

> 166.7 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service