Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1303

Sigma-Aldrich

Trifucosyl-para-lacto-N-hexaose

from human milk, ≥90% (HPLC)

Synonym(s):

α-Fuc-(1→2)-β-Gal-(1→3)-[α-Fuc-(1→4)]-β-GlcNAc-(1→3)-β-Gal-(1→4)-[α-Fuc-(1→3)]-β-GlcNAc-(1→3)-β-Gal-(1→4)-Glc, Leb-Lex-L nonasaccharide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H98N2O43
CAS Number:
Molecular Weight:
1511.39
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

human milk

Assay

≥90% (HPLC)

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]2O[C@@H]3[C@@H](NC(C)=O)[C@@H](O[C@H](CO)[C@H]3O[C@@H]4O[C@@H](C)[C@@H](O)[C@@H](O)[C@@H]4O)O[C@H]5[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]6[C@@H](CO)O[C@@H](O[C@@H]7[C@H](O)[C@@H](O)[C@@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)C(O)O[C@@H]8CO)[C@H](NC(C)=O)[C@H]6O[C@@H]9O[C@@H](C)[C@@H](O)[C@@H](O)[C@@H]9O)[C@@H]5O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C58H98N2O43/c1-12-25(69)30(74)37(81)53(87-12)98-45-22(11-66)94-52(24(60-16(5)68)47(45)101-58-49(33(77)28(72)17(6-61)92-58)103-55-39(83)32(76)27(71)14(3)89-55)102-48-29(73)18(7-62)91-57(41(48)85)99-44-21(10-65)93-51(23(59-15(4)67)46(44)100-54-38(82)31(75)26(70)13(2)88-54)96-43-20(9-64)95-56(40(84)35(43)79)97-42-19(8-63)90-50(86)36(80)34(42)78/h12-14,17-58,61-66,69-86H,6-11H2,1-5H3,(H,59,67)(H,60,68)/t12-,13-,14-,17+,18+,19+,20+,21+,22+,23+,24+,25+,26+,27+,28-,29-,30+,31+,32+,33-,34+,35+,36+,37-,38-,39-,40+,41+,42+,43-,44+,45+,46+,47+,48-,49+,50?,51-,52-,53-,54-,55-,56-,57-,58-/m0/s1

InChI key

UDPJUZHRJSHBSS-SZXKDJIISA-N

Other Notes

Lewis-b, Lewis-X dimer attached to lactose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service