Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1167

Sigma-Aldrich

Tumor Necrosis Factor-α Human Fragment 31-45

≥97% (HPLC)

Synonym(s):

Arg-Arg-Ala-Asn-Ala-Leu-Leu-Ala-Asn-Gly-Val-Glu-Leu-Arg-Asp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C69H122N26O22
CAS Number:
Molecular Weight:
1667.87
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

composition

Peptide content, ~70%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(N)=O)C(=O)NCC(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(O)=O)C(O)=O

Gene Information

human ... TNF(7124)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service