Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T1140

Sigma-Aldrich

Tetramethylammonium perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(ClO4)
CAS Number:
Molecular Weight:
173.60
Beilstein:
3636219
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

SMILES string

C[N+](C)(C)C.[O-]Cl(=O)(=O)=O

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Chen et al.
Bioresource technology, 117, 186-192 (2012-05-23)
The aim of this study was to explore the synergies of laccase, a ligninolytic enzyme, with cellulose and hemicellulase amendments on ensiled corn stover. Molecular signals of lignin decomposition were observed by tetramethylammonium hydroxide thermochemolysis and gas chromatography-mass spectroscopy (TMAH-GC-MS)
Hui Chong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(4), 2091-2098 (2011-11-08)
A new water-soluble conjugated polymer containing fluorene and boron-dipyrromethene repeat units in the backbones (PBF) that exhibits red emission was synthesized and characterized. Cationic PBF forms uniform nanoparticles with negatively charged disodium salt 3,3'-dithiodipropionic acid (SDPA) in aqueous solution through
Yasuyuki Ishida et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 27(10), 1053-1056 (2011-10-12)
Thermochemolysis-gas chromatography in the presence of tetramethylammonium acetate was applied to the direct determination of terephthalic acid (TPA) contained in solid decomposition products obtained from the hydrothermal recycling process of poly(ethylene terephthalate) (PET). On the chromatograms of the hydrothermal decomposition
Marta Matvienko et al.
PloS one, 8(2), e55913-e55913 (2013-02-15)
Several applications of high throughput genome and transcriptome sequencing would benefit from a reduction of the high-copy-number sequences in the libraries being sequenced and analyzed, particularly when applied to species with large genomes. We adapted and analyzed the consequences of
Yusuke Ide et al.
Chemical communications (Cambridge, England), 48(56), 7073-7075 (2012-06-12)
Hiroshima University Silicate-1 (HUS-1), composed of silicate sheets with a halved sodalite cage and the interlayer tetramethylammonium (TMA) cation in the cage, was modified with dimethyldichlorosilane to form the organic derivative in which a dimethyl group was grafted onto the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service