Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0516

Sigma-Aldrich

Thromboxane B2

Synonym(s):

(5Z,9α,11RS,13E,15S)-9,11,15-Trihydroxythromboxa-5,13-dien-1-oic acid, TXB2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O6
CAS Number:
Molecular Weight:
370.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

functional group

carboxylic acid

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\[C@H]1OC(O)C[C@H](O)[C@@H]1C\C=C/CCCC(O)=O

InChI

1S/C20H34O6/c1-2-3-6-9-15(21)12-13-18-16(17(22)14-20(25)26-18)10-7-4-5-8-11-19(23)24/h4,7,12-13,15-18,20-22,25H,2-3,5-6,8-11,14H2,1H3,(H,23,24)/b7-4-,13-12+/t15-,16-,17-,18+,20?/m0/s1

InChI key

XNRNNGPBEPRNAR-JQBLCGNGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Prostaglandin derivative that is released during anaphylaxis; induces arterial contraction and platelet aggregation.
Prostaglandin derivative that is released during anaphylaxis; induces arterial contraction and platelet aggregation. Review of endogenous thromboxane B2 in risk analysis for development of thrombogenic disorders.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Naegele et al.
International journal of cardiology, 224, 226-230 (2016-10-25)
Takotsubo syndrome (TTS) is an acute cardiomyopathy associated with intense physical or emotional stress. The precise mechanisms of the disease remain unclear. The aim of this study was to study alterations in endothelial function, vascular compliance and structure and muscle
Karl Wilhelm Olsson et al.
Pediatric research, 86(3), 333-338 (2018-10-06)
Early identification of infants at risk for complications from patent ductus arteriosus (PDA) may improve treatment outcomes. The aim of this study was to identify biochemical markers associated with persistence of PDA, and with failure of pharmacological treatment for PDA
C R Pace-Asciak et al.
Clinica chimica acta; international journal of clinical chemistry, 246(1-2), 163-182 (1996-03-15)
To test the hypothesis that red wine, by virtue of its relatively high concentration of polyphenols, is more protective against atherosclerosis and coronary heart disease (CHD) than white wine, and that grape juice enriched in one of these, trans-resveratrol, may
Haowen Zhang et al.
European journal of pharmacology, 794, 234-245 (2016-11-30)
High on-aspirin platelet reactivity (HAPR) has been associated with compromised aspirin efficacy in patients with diabetes suffering from acute cardiovascular events, but the key mechanisms remain elusive. The objective of this study was to uncover the potential link between pathogenic
Michal Bijak et al.
BMC complementary and alternative medicine, 17(1), 396-396 (2017-08-12)
Arachidonic acid metabolism by cyclooxygenase (COX) is a major pathway for blood platelets' activation, which is associated with pro-thrombotic platelet activity and the production of pro-inflammatory mediators. Inhibition of COX activity is one of the major means of anti-platelet pharmacotherapy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service