Skip to Content
MilliporeSigma
All Photos(1)

Documents

T0386

Sigma-Aldrich

3,3′,5-Triiodothyropropionic acid

98-99%

Synonym(s):

3-(4-[4-Hydroxy-3-iodophenoxy]-3,5-diiodophenyl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11I3O4
CAS Number:
Molecular Weight:
635.96
MDL number:
UNSPSC Code:
12352200

Assay

98-99%

storage temp.

−20°C

SMILES string

OC(=O)CCc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Naidoo
Analytical biochemistry, 91(2), 543-547 (1978-12-01)
A thin-layer chromatographic method is described for the separation of the acetic and propionic acid analogs of thyroxine and triiodothyronine, which are inseparable by currently available methods. The separation is achieved on silica gel H in a solvent system consisting
T Takeda et al.
The Journal of clinical endocrinology and metabolism, 80(7), 2033-2040 (1995-07-01)
3,5,3,'-Triiodothyroacetic acid (Triac) has been used in therapy of resistance to thyroid hormone on an empirical basis and appears beneficial in some studies. We observed that the T3 analogs, Triac and 3,5,3'-triiodothyropropionic acid (Triprop), have a higher affinity for the
T Yamamoto et al.
Journal of biochemistry, 128(1), 121-128 (2000-07-06)
The NADP(H)-linked oxidoreductase activity of a major isozyme of human liver 3 alpha-hydroxysteroid dehydrogenase was activated 5-, 4-, and 2-fold by D-thyroxine (T(4)), L-T(4) and DL-3,3', 5'-triiodothyronine (reverse T(3)), respectively. Kinetic analysis of the activation indicated that D-T(4), L-T(4), and
S B Hoath et al.
American journal of diseases of children (1960), 138(3), 251-253 (1984-03-01)
We have shown a positive correlation between the epidermal growth factor concentration in neonatal mouse skin and prior treatment with thyroxine (T4) or triiodothyronine (T3). The results of the present study indicate similar activity of other levorotatory iodothyronine analogues and
V Cody
Endocrine research, 14(2-3), 165-176 (1988-01-01)
The crystal and molecular structures of 3,5,3'-triiodothyropropionic acid (T3P), determined as an N-diethanolamine salt, were carried out and the results are compared with those of other thyroid hormone structures. These data show that T3P has an unusual conformation with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service