Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0158

Sigma-Aldrich

Calcitonin porcine

≥97% (HPLC)

Synonym(s):

Thyrocalcitonin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C159H232N46O45S3
CAS Number:
Molecular Weight:
3604.02
EC Number:
MDL number:
UNSPSC Code:
51111800

Assay

≥97% (HPLC)

storage temp.

−20°C

Amino Acid Sequence

Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Ser-Ala-Tyr-Trp-Arg-Asn-Leu-Asn-Asn-Phe-His-Arg-Phe-Ser-Gly-Met-Gly-Phe-Gly-Pro-Glu-Thr-Pro-NH2 [Disulfide Bridge: 1-7]

Biochem/physiol Actions

Hypocalcemic hormone produced by the parafollicular C cells of the thyroid or by the ultimobranchial bodies of nonmammalian vertebrates. Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes.

Other Notes

32 amino acid polypeptide, 8 of which are conserved across all species.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service